Anti-CD2 Rat Monoclonal Antibody [clone: 377830]

Supplier: R & D Systems
MAB2828
220020-121EA 594.37 USD
220020-121
Anti-CD2 Rat Monoclonal Antibody [clone: 377830]
Antibodies
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: 377830
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR