Anti-ESAM Rat Monoclonal Antibody [clone: 340212]

Supplier: R & D Systems
MAB2827
220020-119EA 566.89 USD
220020-119
Anti-ESAM Rat Monoclonal Antibody [clone: 340212]
Antibodies
Type: Primary
Antigen: ESAM
Clonality: Monoclonal
Clone: 340212
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR