Anti-IL31 Mouse Monoclonal Antibody [clone: 308227]

Supplier: R & D Systems
MAB2824
220020-117EA 594.37 USD
220020-117
Anti-IL31 Mouse Monoclonal Antibody [clone: 308227]
Antibodies
Type: Primary
Antigen: IL31
Clonality: Monoclonal
Clone: 308227
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR