Anti-EGR1 Rat Monoclonal Antibody [clone: 406913]

Supplier: R & D Systems
MAB2818
220020-115EA 594.37 USD
220020-115
Anti-EGR1 Rat Monoclonal Antibody [clone: 406913]
Antibodies
Type: Primary
Antigen: EGR1
Clonality: Monoclonal
Clone: 406913
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR