Anti-CCL2 Mouse Monoclonal Antibody [clone: 280702]

Supplier: R & D Systems
MAB28171
220020-114EA 763.57 USD
220020-114
Anti-CCL2 Mouse Monoclonal Antibody [clone: 280702]
Antibodies
Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone: 280702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR