Anti-CCR10 Rat Monoclonal Antibody [clone: 248918]

Supplier: R & D Systems
MAB2815
220020-112EA 594.37 USD
220020-112
Anti-CCR10 Rat Monoclonal Antibody [clone: 248918]
Antibodies
Type: Primary
Antigen: CCR10
Clonality: Monoclonal
Clone: 248918
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR