Anti-GLP1R Mouse Monoclonal Antibody [clone: 419208]

Supplier: R & D Systems
MAB28141
220020-111EA 639.2 USD
220020-111
Anti-GLP1R Mouse Monoclonal Antibody [clone: 419208]
Antibodies
Type: Primary
Antigen Symbol: GLP1R
Antigen Name: glucagon-like peptide 1 receptor
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 419208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR