Anti-GLP1R Mouse Monoclonal Antibody [clone: 197920]

Supplier: R & D Systems
MAB2814
220020-110EA 655.11 USD
220020-110
Anti-GLP1R Mouse Monoclonal Antibody [clone: 197920]
Antibodies
Type: Primary
Antigen Symbol: GLP1R
Antigen Name: glucagon-like peptide 1 receptor
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 197920
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR