Anti-CDX4 Mouse Monoclonal Antibody [clone: 322007]

Supplier: R & D Systems
MAB2807
220020-108EA 561.11 USD
220020-108
Anti-CDX4 Mouse Monoclonal Antibody [clone: 322007]
Antibodies
Type: Primary
Antigen: CDX4
Clonality: Monoclonal
Clone: 322007
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR