Anti-CLEC4D Mouse Monoclonal Antibody [clone: 413512]

Supplier: R & D Systems
MAB2806
220020-107EA 577.02 USD
220020-107
Anti-CLEC4D Mouse Monoclonal Antibody [clone: 413512]
Antibodies
Type: Primary
Antigen: CLEC4D
Clonality: Monoclonal
Clone: 413512
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR