Anti-RLN2 Rat Monoclonal Antibody [clone: 349102]

Supplier: R & D Systems
MAB2804
220020-105EA 660.89 USD
220020-105
Anti-RLN2 Rat Monoclonal Antibody [clone: 349102]
Antibodies
Type: Primary
Antigen: RLN2
Clonality: Monoclonal
Clone: 349102
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR