Anti-IL1RN Mouse Monoclonal Antibody [clone: 10309]

Supplier: R & D Systems
MAB280
220020-104EA 721.63 USD
220020-104
Anti-IL1RN Mouse Monoclonal Antibody [clone: 10309]
Antibodies
Type: Primary
Antigen: IL1RN
Clonality: Monoclonal
Clone: 10309
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR