Anti-Sleeping Beauty Transposase Mouse Monoclonal Antibody [clone: 324622]

Supplier: R & D Systems
MAB2798
220020-103EA 594.37 USD
220020-103
Anti-Sleeping Beauty Transposase Mouse Monoclonal Antibody [clone: 324622]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: Sleeping Beauty Transposase
Antigen Synonyms:
Clonality: Monoclonal
Clone: 324622
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR