Anti-CRHBP Mouse Monoclonal Antibody [clone: 395108]

Supplier: R & D Systems
MAB2796
220020-102EA 561.11 USD
220020-102
Anti-CRHBP Mouse Monoclonal Antibody [clone: 395108]
Antibodies
Type: Primary
Antigen: CRHBP
Clonality: Monoclonal
Clone: 395108
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR