Anti-FLRT1 Mouse Monoclonal Antibody [clone: 357721]

Supplier: R & D Systems
MAB2794
220020-100EA 577.02 USD
220020-100
Anti-FLRT1 Mouse Monoclonal Antibody [clone: 357721]
Antibodies
Type: Primary
Antigen Symbol: FLRT1
Antigen Name: fibronectin leucine rich transmembrane protein 1
Antigen Synonyms: SPG68
Clonality: Monoclonal
Clone: 357721
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR