Anti-SCGB3A1 Mouse Monoclonal Antibody [clone: 410221]

Supplier: R & D Systems
MAB2790
220020-097EA 561.11 USD
220020-097
Anti-SCGB3A1 Mouse Monoclonal Antibody [clone: 410221]
Antibodies
Type: Primary
Antigen: SCGB3A1
Clonality: Monoclonal
Clone: 410221
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR