Anti-RELM gamma Mouse Monoclonal Antibody [clone: 395310]

Supplier: R & D Systems
MAB2789
220020-095EA 584.25 USD
220020-095
Anti-RELM gamma Mouse Monoclonal Antibody [clone: 395310]
Antibodies
Type: Primary
Antigen: RELM gamma
Clonality: Monoclonal
Clone: 395310
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR