Anti-CLEC4F Rat Monoclonal Antibody [clone: 370901]

Supplier: R & D Systems
MAB2784
220020-093EA 623.29 USD
220020-093
Anti-CLEC4F Rat Monoclonal Antibody [clone: 370901]
Antibodies
Type: Primary
Antigen: CLEC4F
Clonality: Monoclonal
Clone: 370901
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR