Anti-SDC1 Rat Monoclonal Antibody [clone: 359103]

Supplier: R & D Systems
MAB2780
220020-091EA 655.11 USD
220020-091
Anti-SDC1 Rat Monoclonal Antibody [clone: 359103]
Antibodies
Type: Primary
Antigen: SDC1
Clonality: Monoclonal
Clone: 359103
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR