Anti-RTN4RL2 Mouse Monoclonal Antibody [clone: 333413]

Supplier: R & D Systems
MAB2776
220020-087EA 577.02 USD
220020-087
Anti-RTN4RL2 Mouse Monoclonal Antibody [clone: 333413]
Antibodies
Type: Primary
Antigen: RTN4RL2
Clonality: Monoclonal
Clone: 333413
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR