Anti-CD300LF Rat Monoclonal Antibody [clone: 234903]

Supplier: R & D Systems
MAB2774
220020-086EA 616.06 USD
220020-086
Anti-CD300LF Rat Monoclonal Antibody [clone: 234903]
Antibodies
Type: Primary
Antigen: CD300LF
Clonality: Monoclonal
Clone: 234903
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR