Anti-IL22RA1 Mouse Monoclonal Antibody [clone: 305405]

Supplier: R & D Systems
MAB2770
220020-084EA 639.2 USD
220020-084
Anti-IL22RA1 Mouse Monoclonal Antibody [clone: 305405]
Antibodies
Type: Primary
Antigen: IL22RA1
Clonality: Monoclonal
Clone: 305405
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR