Anti-SOX7 Mouse Monoclonal Antibody [clone: 288610]

Supplier: R & D Systems
MAB2766
220020-081EA 577.02 USD
220020-081
Anti-SOX7 Mouse Monoclonal Antibody [clone: 288610]
Antibodies
Type: Primary
Antigen: SOX7
Clonality: Monoclonal
Clone: 288610
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR