Anti-VWF Mouse Monoclonal Antibody [clone: 490628]

Supplier: R & D Systems
MAB27642
220020-080EA 639.2 USD
220020-080
Anti-VWF Mouse Monoclonal Antibody [clone: 490628]
Antibodies
Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: 490628
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR