Anti-VWF Mouse Monoclonal Antibody [clone: 210909]

Supplier: R & D Systems
MAB2764
220020-078EA 779.48 USD
220020-078
Anti-VWF Mouse Monoclonal Antibody [clone: 210909]
Antibodies
Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: 210909
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR