Anti-CXCL1/2/3 Mouse Monoclonal Antibody [clone: 29702]

Supplier: R & D Systems
MAB2761
220020-077EA 577.02 USD
220020-077
Anti-CXCL1/2/3 Mouse Monoclonal Antibody [clone: 29702]
Antibodies
Type: Primary
Antigen: CXCL1/2/3
Clonality: Monoclonal
Clone: 29702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR