Anti-TLR4 Rat Monoclonal Antibody [clone: 267518]

Supplier: R & D Systems
MAB2759
220020-074EA 633.42 USD
220020-074
Anti-TLR4 Rat Monoclonal Antibody [clone: 267518]
Antibodies
Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: 267518
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR