Anti-CLEC4B1 Rat Monoclonal Antibody [clone: 349214]

Supplier: R & D Systems
MAB2757
220020-073EA 577.02 USD
220020-073
Anti-CLEC4B1 Rat Monoclonal Antibody [clone: 349214]
Antibodies
Type: Primary
Antigen: CLEC4B1
Clonality: Monoclonal
Clone: 349214
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR