Anti-PIRB Rat Monoclonal Antibody [clone: 326414]

Supplier: R & D Systems
MAB2754
220020-071EA 647.88 USD
220020-071
Anti-PIRB Rat Monoclonal Antibody [clone: 326414]
Antibodies
Type: Primary
Antigen: PIRB
Clonality: Monoclonal
Clone: 326414
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR