Anti-FABP9 Rat Monoclonal Antibody [clone: 324926]

Supplier: R & D Systems
MAB2750
220020-070EA 561.11 USD
220020-070
Anti-FABP9 Rat Monoclonal Antibody [clone: 324926]
Antibodies
Type: Primary
Antigen Symbol: FABP9
Antigen Name: fatty acid binding protein 9, testis
Antigen Synonyms: 1700007P10Rik, Perf, Perf15, Tlbp
Clonality: Monoclonal
Clone: 324926
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR