Anti-PROCR Rat Monoclonal Antibody [clone: 432732]

Supplier: R & D Systems
MAB27491
220020-068EA 561.11 USD
220020-068
Anti-PROCR Rat Monoclonal Antibody [clone: 432732]
Antibodies
Type: Primary
Antigen: PROCR
Clonality: Monoclonal
Clone: 432732
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR