Anti-PROCR Rat Monoclonal Antibody [clone: 432714]

Supplier: R & D Systems
MAB2749
220020-067EA 561.11 USD
220020-067
Anti-PROCR Rat Monoclonal Antibody [clone: 432714]
Antibodies
Type: Primary
Antigen: PROCR
Clonality: Monoclonal
Clone: 432714
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR