Anti-NTNG2 Rat Monoclonal Antibody [clone: 349724]

Supplier: R & D Systems
MAB27441
220020-065EA 577.02 USD
220020-065
Anti-NTNG2 Rat Monoclonal Antibody [clone: 349724]
Antibodies
Type: Primary
Antigen: NTNG2
Clonality: Monoclonal
Clone: 349724
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR