Anti-IL10RA Mouse Monoclonal Antibody [clone: 714212]

Supplier: R & D Systems
MAB2742
102885-498EA 600.15 USD
102885-498
Anti-IL10RA Mouse Monoclonal Antibody [clone: 714212]
Antibodies
Type: Primary
Antigen: IL10RA
Clonality: Monoclonal
Clone: 714212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR