Anti-ITGB1BP2 Mouse Monoclonal Antibody [clone: 308705]

Supplier: R & D Systems
MAB2740
220020-063EA 561.11 USD
220020-063
Anti-ITGB1BP2 Mouse Monoclonal Antibody [clone: 308705]
Antibodies
Type: Primary
Antigen: ITGB1BP2
Clonality: Monoclonal
Clone: 308705
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR