Anti-CFB Mouse Monoclonal Antibody [clone: 313011]

Supplier: R & D Systems
MAB2739
220020-061EA 582.8 USD
220020-061
Anti-CFB Mouse Monoclonal Antibody [clone: 313011]
Antibodies
Type: Primary
Antigen: CFB
Clonality: Monoclonal
Clone: 313011
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR