Anti-CD70 Mouse Monoclonal Antibody [clone: 301731]

Supplier: R & D Systems
MAB2738
220020-060EA 655.11 USD
220020-060
Anti-CD70 Mouse Monoclonal Antibody [clone: 301731]
Antibodies
Type: Primary
Antigen: CD70
Clonality: Monoclonal
Clone: 301731
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR