Anti-SDC3 Rat Monoclonal Antibody [clone: 312607]

Supplier: R & D Systems
MAB2734
220020-058EA 594.37 USD
220020-058
Anti-SDC3 Rat Monoclonal Antibody [clone: 312607]
Antibodies
Type: Primary
Antigen: SDC3
Clonality: Monoclonal
Clone: 312607
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR