Anti-FGF9 Mouse Monoclonal Antibody [clone: 36923]

Supplier: R & D Systems
MAB2731
220020-057EA 772.25 USD
220020-057
Anti-FGF9 Mouse Monoclonal Antibody [clone: 36923]
Antibodies
Type: Primary
Antigen Symbol: FGF9
Antigen Name: fibroblast growth factor 9
Antigen Synonyms: FGF-9, GAF, HBFG-9, HBGF-9, SYNS3
Clonality: Monoclonal
Clone: 36923
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR