Anti-LMO2 Mouse Monoclonal Antibody [clone: 313606]

Supplier: R & D Systems
MAB2726
102885-496EA 561.11 USD
102885-496
Anti-LMO2 Mouse Monoclonal Antibody [clone: 313606]
Antibodies
Type: Primary
Antigen: LMO2
Clonality: Monoclonal
Clone: 313606
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR