Anti-CD200 Mouse Monoclonal Antibody [clone: 325516]

Supplier: R & D Systems
MAB27241
220020-053EA 594.37 USD
220020-053
Anti-CD200 Mouse Monoclonal Antibody [clone: 325516]
Antibodies
Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone: 325516
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR