Anti-CD200 Mouse Monoclonal Antibody [clone: 325520]

Supplier: R & D Systems
MAB2724
220020-052EA 594.37 USD
220020-052
Anti-CD200 Mouse Monoclonal Antibody [clone: 325520]
Antibodies
Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone: 325520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR