Anti-PIGR Mouse Monoclonal Antibody [clone: 825732]

Supplier: R & D Systems
MAB27171
102885-494EA 594.37 USD
102885-494
Anti-PIGR Mouse Monoclonal Antibody [clone: 825732]
Antibodies
Type: Primary
Antigen: PIGR
Clonality: Monoclonal
Clone: 825732
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR