Anti-PIGR Mouse Monoclonal Antibody [clone: 825724]

Supplier: R & D Systems
MAB2717
102885-492EA 594.37 USD
102885-492
Anti-PIGR Mouse Monoclonal Antibody [clone: 825724]
Antibodies
Simple Western

Type: Primary
Antigen: PIGR
Clonality: Monoclonal
Clone: 825724
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR