Anti-CDH6 Mouse Monoclonal Antibody [clone: 427909]

Supplier: R & D Systems
MAB2715
220020-049EA 623.29 USD
220020-049
Anti-CDH6 Mouse Monoclonal Antibody [clone: 427909]
Antibodies
Type: Primary
Antigen: CDH6
Clonality: Monoclonal
Clone: 427909
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR