Anti-IL1F5 Rat Monoclonal Antibody [clone: 759207]

Supplier: R & D Systems
MAB2714
102885-490EA 594.37 USD
102885-490
Anti-IL1F5 Rat Monoclonal Antibody [clone: 759207]
Antibodies
Type: Primary
Antigen: IL1F5
Clonality: Monoclonal
Clone: 759207
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR