Anti-CTF2 Rat Monoclonal Antibody [clone: 321525]

Supplier: R & D Systems
MAB2709
220020-046EA 561.11 USD
220020-046
Anti-CTF2 Rat Monoclonal Antibody [clone: 321525]
Antibodies
Type: Primary
Antigen: CTF2
Clonality: Monoclonal
Clone: 321525
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR