Anti-MSR1 Mouse Monoclonal Antibody [clone: 351620]

Supplier: R & D Systems
MAB27081
220020-045EA 655.11 USD
220020-045
Anti-MSR1 Mouse Monoclonal Antibody [clone: 351620]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: MSR1
Clonality: Monoclonal
Clone: 351620
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR