Anti-CD300E Rat Monoclonal Antibody [clone: 233810]

Supplier: R & D Systems
MAB2705
220020-043EA 594.37 USD
220020-043
Anti-CD300E Rat Monoclonal Antibody [clone: 233810]
Antibodies
Type: Primary
Antigen: CD300E
Clonality: Monoclonal
Clone: 233810
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR