Anti-ACY1 Rat Monoclonal Antibody [clone: 47573]

Supplier: R & D Systems
MAB2700
220020-041EA 561.11 USD
220020-041
Anti-ACY1 Rat Monoclonal Antibody [clone: 47573]
Antibodies
Type: Primary
Antigen: ACY1
Clonality: Monoclonal
Clone: 47573
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR